missing translation for 'onlineSavingsMsg'
Learn More

Pentamethylbenzene, 99%

Product Code. 11396816
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11396816 5 g 5g
15408829 25 g 25g
11306826 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11396816 Supplier Thermo Scientific Alfa Aesar Supplier No. L04163.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 700-12-9 | C11H16 | 148.249 g/mol

Pentamethylbenzene is used to prepare a mixture of nitropentamethylbenzene and 2,3,4,5-tetramethylbenzyl nitrate1. It was also used to prepare propene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pentamethylbenzene is used to prepare a mixture of nitropentamethylbenzene and 2,3,4,5-tetramethylbenzyl nitrate1. It was also used to prepare propene.

Notes
Incompatible with oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 700-12-9
Molecular Formula C11H16
Molecular Weight (g/mol) 148.249
MDL Number MFCD00008522
InChI Key BEZDDPMMPIDMGJ-UHFFFAOYSA-N
Synonym pentamethylbenzene, benzene, pentamethyl, pentamethylbenzol, benzene, 1,2,3,4,5-pentamethyl, pentamethyl phenyl, acmc-1bkt8, ksc492e3d, 1,3,4,5-pentamethylbenzene, pentamethylbenzene 5g
PubChem CID 12784
ChEBI CHEBI:38998
IUPAC Name 1,2,3,4,5-pentamethylbenzene
SMILES CC1=CC(=C(C(=C1C)C)C)C

Specifications

Melting Point 50°C to 51°C
Density 0.917
Boiling Point 231°C to 232°C
Flash Point 91°C (195°F)
Quantity 5 g
UN Number UN1325
Beilstein 1905349
Formula Weight 148.25
Percent Purity 99%
Chemical Name or Material Pentamethylbenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.