missing translation for 'onlineSavingsMsg'
Learn More

Pentamethylbenzonitrile, 98%

Product Code. 11419120
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11419120 5 g 5g
11429120 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 11419120

Brand: Thermo Scientific Alfa Aesar A14733.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 5144-10-5 | C5H8N2O | 112.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5144-10-5
MDL Number MFCD00015450

Specifications

Melting Point 158°C to 160°C
Molecular Formula C12H15N
Quantity 5 g
UN Number UN3439
Synonym pentamethylbenzonitrile, 6-aminobutyric acid, pentamethylbenzonitril, acmc-1axcs, benzonitrile,2,3,4,5,6-pentamethyl, 2,3,4,5,6-pentamethylbenzenecarbonitrile
InChI Key NCDPYDGGUAKABA-UHFFFAOYSA-N
SMILES CC1=C(C(=C(C(=C1C)C)C#N)C)C
IUPAC Name 2,3,4,5,6-pentamethylbenzonitrile
Molecular Weight (g/mol) 173.26
PubChem CID 78824
Formula Weight 173.26
Percent Purity 98%
Chemical Name or Material Pentamethylbenzonitrile
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.