missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Pentamidine isethionate salt, 97%

Product Code. 15891022 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15891022 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15891022 Supplier Thermo Scientific Chemicals Supplier No. 461860010
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 140-64-7 | C19H24N4O2·2C2H6O4S | 592.68 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 140-64-7
Melting Point 188°C to 194°C
Boiling Point 539.4°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Molecular Formula C19H24N4O2·2C2H6O4S
Quantity 1 g
Solubility Information Solubility in water: soluble. Other solubilities: soluble in glycerol,slightly soluble in alcohol,insoluble in ether,acetone,chloroform
InChI Key YBVNFKZSMZGRAD-UHFFFAOYSA-N
Molecular Weight (g/mol) 592.68
PubChem CID 8813
ChEBI CHEBI:7977
Formula Weight 592.68
Percent Purity 97%
Chemical Name or Material Pentamidine isethionate salt
Show More Show Less
compliance-icons
Product Identifier
  • Pentamidine isethionate salt, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Pentamidine isethionate salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.