missing translation for 'onlineSavingsMsg'
Learn More

Pentan-3-One, Technical, Fisher Chemical

Product Code. 11463673 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
250 mL
Packaging:
Amber glass bottle
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11463673 250 mL Amber glass bottle 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11463673 Supplier Fisher Chemical Supplier No. P/1100/07
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 96-22-0
Melting Point -40°C
Color Colorless
pH 6.2
Boiling Point 100°C
Packaging Amber glass bottle
Molecular Formula C5H10O
MDL Number MFCD00009320
Quantity 250 mL
Synonym 3-pentanone, diethyl ketone, propione, ethyl ketone, dimethylacetone, metacetone, methacetone, diethylcetone, ethyl propionyl, diethylketone
InChI Key FDPIMTJIUBPUKL-UHFFFAOYSA-N
SMILES CCC(=O)CC
IUPAC Name pentan-3-one
Molecular Weight (g/mol) 86.13
PubChem CID 7288
Vapor Density 3
ChEBI CHEBI:67886
Formula Weight 86.13g/mol
Physical Form Liquid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Pentan-3-one
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H336-May cause drowsiness or dizziness.
  • H335-May cause respiratory irritation.
  • H319-Causes serious eye irritation.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.