missing translation for 'onlineSavingsMsg'
Learn More

Pentane, Laboratory Reagent, ≥95.0% (GC), Solstice

Product Code. 15651490 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
4 x 5 L
Packaging:
Aluminum Bottle
Unit Size:
Pack of 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15651490 4 x 5 L Aluminum Bottle Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 15651490 Supplier Honeywell Research Chemicals Supplier No. 156074X5L
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Laboratory Reagent,≥95.0% (GC)

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Specifications

CAS 109-66-0
Density 0.626g/mL at 25°C
Quantity 4 x 5 L
Explosion Limit 0.083
Flash Point −49°C (−56.2°F)
Ignition Point 500°F
Packaging Aluminum Bottle
Molecular Formula C5H12
Linear Formula CH3(CH2)3CH3
MDL Number MFCD00009498
UN Number UN1265
Beilstein 969132
Synonym n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
InChI Key OFBQJSOFQDEBGM-UHFFFAOYSA-N
SMILES CCCCC
IUPAC Name pentane
Molecular Weight (g/mol) 72.15
PubChem CID 8003
ChEBI CHEBI:37830
Formula Weight 72.15g/mol
Percent Purity ≥95.0% (GC)
Grade Laboratory
Chemical Name or Material Pentane
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Pentane
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H336-May cause drowsiness or dizziness.
  • H411-Toxic to aquatic life with long lasting effects.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Pentane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.