missing translation for 'onlineSavingsMsg'
Learn More

Pentane, AMD CHROMASOLV™, ≥99%, Solstice

Product Code. 15644690
Change view
Click to view available options
Quantity:
2.5 L
4 x 2.5 L
Packaging:
Glass Bottle
Unit Size:
2.5 L
Pack of 4
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15644690 4 x 2.5 L Glass Bottle Pack of 4
15634690 2.5 L Glass Bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 15644690 Supplier Honeywell-Chromasolv Supplier No. 348944X2.5L
Solstice Chromasolv Promotion
Use promo code 27944
View all promotions
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

CHROMASOLV™ AMD,≥99%

  • Solvents designed for high-performance thin-layer chromatography
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-66-0
Molecular Formula C5H12
Molecular Weight (g/mol) 72.15
MDL Number MFCD00009498
InChI Key OFBQJSOFQDEBGM-UHFFFAOYSA-N
Synonym n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
PubChem CID 8003
ChEBI CHEBI:37830
IUPAC Name pentane
SMILES CCCCC

Specifications

Melting Point -130°C (literature)
CAS Max % ≤100.0000%
Density 0.626 g/mL at 25 °C(lit.)
Quantity 4 x 2.5 L
Flash Point <−35°C
Packaging Glass Bottle
Linear Formula CH3(CH2)3CH3
UN Number UN1265
Beilstein 969132
Boiling Point 36°C to 37°C
Formula Weight 72.15
Percent Purity >99%
Grade High Purity
Chemical Name or Material Pentane, AMD CHROMASOLV™, ≥99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.