missing translation for 'onlineSavingsMsg'
Learn More

Pentane, CHROMASOLV™, for residue analysis (of high-volatile halogenated hydrocarbons), Solstice

Product Code. 15677040
Change view
Click to view available options
Quantity:
1 L
2.5 L
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15677040 1 L 1 L
15687040 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 15677040 Supplier Honeywell-Chromasolv Supplier No. 768661L
Solstice Chromasolv Promotion
Use promo code 27944
View all promotions
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

CHROMASOLV™, for residue analysis (of high-volatile halogenated hydrocarbons),≥99.0%

  • For gas chromatography (GC)
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
  • Non-volatile matter and water content are specified for each solvent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-66-0
Molecular Formula C5H12
Molecular Weight (g/mol) 72.15
MDL Number MFCD00009498
InChI Key OFBQJSOFQDEBGM-UHFFFAOYSA-N
Synonym n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
PubChem CID 8003
ChEBI CHEBI:37830
IUPAC Name pentane
SMILES CCCCC

Specifications

Density 0.626g/mL at 25°C
Quantity 1 L
Explosion Limit 0.083
Flash Point −49°C (−56.2°F)
Ignition Point 500°F
Linear Formula CH3(CH2)3CH3
UN Number UN1265
Beilstein 969132
Formula Weight 72.15g/mol
Percent Purity ≥99%, ≥99% (GC)
Grade CHROMASOLV
Chemical Name or Material Pentane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.