missing translation for 'onlineSavingsMsg'
Learn More

Pentanenitrile-d9, TRC

Product Code. 30457012
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30457012 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30457012 Supplier Toronto Research Chemicals Supplier No. TRCV0914915MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Spécification

Chemical Name or Material Pentanenitrile-d9
CAS 1030939-70-8
Molecular Formula C5 D9 N
Quantity 5 mg
Synonym Pentanenitrile-2,2,3,3,4,4,5,5,5-d9 (ACI), Perdeuterated-1-cyanobutane
InChI Formula InChI=1S/C5H9N/c1-2-3-4-5-6/h2-4H2,1H3/i1D3,2D2,3D2,4D2
SMILES [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C#N
IUPAC Name 2,2,3,3,4,4,5,5,5-nonadeuteriopentanenitrile
Molecular Weight (g/mol) 92.19
Formula Weight 92.13
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.