missing translation for 'onlineSavingsMsg'
Learn More

Pentanoic-3,3-d2 Acid, TRC

Product Code. 30457114
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30457114 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30457114 Supplier Toronto Research Chemicals Supplier No. TRCV091421100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pentanoic-3,3-D2 Acid
CAS 83741-74-6
Molecular Formula C5 D2 H8 O2
Quantity 100 mg
Synonym Pentanoic-3,3-d2 acid (9CI','ACI), β-Dideuteropentanoic acid, Pentanoic acid-3,3-d2, Valeric acid-d2, Valeric-β,β-d2 acid, Pentanoic-3,3-D2 acid
InChI Formula InChI=1S/C5H10O2/c1-2-3-4-5(6)7/h2-4H2,1H3,(H,6,7)/i3D2
SMILES [2H]C([2H])(CC)CC(=O)O
IUPAC Name 3,3-dideuteriopentanoic acid
Molecular Weight (g/mol) 104.144
Formula Weight 104.081
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.