missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentanoyl-d9 chloride, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.5g
Specifications
Specifications
| Chemical Name or Material | Valeryl-d9 Chloride |
| CAS | 148587-09-1 |
| Molecular Formula | C5 D9 Cl O |
| Quantity | 0.5 g |
| Synonym | Pentanoyl-d9 chloride (9 CI), Valeryl-d9 Chloride, Pentanoyl Chloride-D9, Pentanoic acid chloride-d9 |
| InChI Formula | InChI=1 S/C5H9ClO/c1-2-3-4-5(6)7/h2-4H2,1H3/i1D3,2D2,3D2,4D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C(=O)Cl |
| IUPAC Name | 2,2,3,3,4,4,5,5,5-nonadeuteriopentanoyl chloride |
| Molecular Weight (g/mol) | 129.6328 |
| Formula Weight | 129.0907 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction