missing translation for 'onlineSavingsMsg'
Learn More

Pentaphenylcyclopentadiene, 99%

Product Code. 11332219
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11332219 1 g 1g
15453637 250 mg 250mg
11342219 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11332219 Supplier Thermo Scientific Alfa Aesar Supplier No. H27223.03
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 2519-10-0 | C35H26 | 446.593 g/mol

1,2,3,4,5-Pentaphenyl-1,3-cyclopentadiene (PPCP) was used as dopant for poly(3-alkylthiophene) diodes to enhance their electroluminescence efficiency, in the preparation of monodisperse and highly fluorescent silica nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,3,4,5-Pentaphenyl-1,3-cyclopentadiene (PPCP) was used as dopant for poly(3-alkylthiophene) diodes to enhance their electroluminescence efficiency, in the preparation of monodisperse and highly fluorescent silica nanoparticles.

Solubility
Soluble in pyridine 5 mg/ml.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2519-10-0
Molecular Formula C35H26
Molecular Weight (g/mol) 446.593
MDL Number MFCD09909572
InChI Key YGLVWOUNCXBPJF-UHFFFAOYSA-N
PubChem CID 635023
IUPAC Name (2,3,4,5-tetraphenylcyclopenta-1,4-dien-1-yl)benzene
SMILES C1=CC=C(C=C1)C2C(=C(C(=C2C3=CC=CC=C3)C4=CC=CC=C4)C5=CC=CC=C5)C6=CC=CC=C6

Specifications

Melting Point 254°C to 257°C
Quantity 1 g
Synonym 1,2,3,4,5-pentaphenyl-1,3-cyclopentadiene, pentaphenylcyclopentadiene, cyclopenta-1,3-diene-1,2,3,4,5-pentaylpentabenzene, 2,3,4,5-tetraphenylcyclopenta-1,4-dien-1-yl benzene, benzene,1,1',1,1',1-1,3-cyclopentadiene-1,2,3,4,5-pentayl pentakis, 2,3,4,5-tetraphenylcyclopenta-2,4-dien-1-yl benzene, acmc-1cek1, 2,3,4,5-tetraphenyl-1-cyclopenta-1,4-dienyl benzene, benzene, 1,1',1,1',1-1,3-cyclopentadiene-1,2,3,4,5-pentayl pentakis, 2,3,4,5-tetraphenylcyclopenta-2,4-dienyl benzene
Solubility Information Soluble in pyridine 5mg/ml.
Formula Weight 446.59
Percent Purity 99%
Chemical Name or Material Pentaphenylcyclopentadiene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.