missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentoxifylline, MedChemExpress
Description
Pentoxifylline (BL-191), a haemorheological agent, is an orally active non-selective phosphodiesterase (PDE) inhibitor, with immune modulation, anti-inflammatory, hemorheological, anti-fibrinolytic and anti-proliferation effects. Pentoxifylline can be used for the research of peripheral vascular disease, cerebrovascular disease and a number of other conditions involving a defective regional microcirculation.
Specifications
Specifications
| Chemical Name or Material | Pentoxifylline |
| CAS | 5-6-6493 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C13H18N4O3 |
| Quantity | 1 g |
| Synonym | BL-191 PTX Oxpentifylline |
| Solubility Information | H2O : 93.3 mg/mL (335.24 mM; Need ultrasonic and warming) ∣DMSO : ≥ 2.8 mg/mL (10.06 mM) |
| SMILES | O=C(N1CCCCC(C)=O)N(C)C2=C(N(C)C=N2)C1=O |
| Molecular Weight (g/mol) | 278.31 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?