missing translation for 'onlineSavingsMsg'
Learn More

Pentyl isothiocyanate, 95+%, Thermo Scientific™

Product Code. 10362285
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10362285 5 g 5g
10436682 10 g 10g
10011894 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10362285 Supplier Thermo Scientific Maybridge Supplier No. TL04631DE
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 629-12-9
Molecular Formula C6H11NS
Molecular Weight (g/mol) 129.221
InChI Key SGHJUJBYMSVAJY-UHFFFAOYSA-N
Synonym pentyl isothiocyanate, n-amyl isothiocyanate, 1-pentyl isothiocyanate, amyl isothiocyanate, n-pentyl isothiocyanate, pentylisothiocyanate, unii-kl6u8nl15r, n-amylisothiocyanate, 1-isothiocyanato-pentane, pentane,1-isothiocyanato
PubChem CID 69415
IUPAC Name 1-isothiocyanatopentane
SMILES CCCCCN=C=S

Specifications

Density 0.93g/mL
Assay Percent Range 95%
Linear Formula CH3(CH2)4NCS
Quantity 5 g
Formula Weight 129.23
Percent Purity 95%
Chemical Name or Material Pentyl isothiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.