missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Pentylenetetrazole

Product Code. 18722594
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18722594 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18722594 Supplier R&D Systems™ Supplier No. 2687/1G
  • ONLINE EXCLUSIVE PRICE
    142.21 €
    127.05 € / 1g
    Save 15.16 €
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. CNS stimulant

CNS stimulant that induces kindling in vivo. Causes alterations in excitatory and inhibitory neurotransmitter systems.
TRUSTED_SUSTAINABILITY

Specifications

CAS 54-95-5
Quantity 1 g
Product Line Tocris
Molecular Formula C6H10N4
Formula Weight 138.17
Purity >99%
Molecular Weight (g/mol) 138.174
Chemical Name or Material Pentylenetetrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.