missing translation for 'onlineSavingsMsg'
Learn More

Pentylmagnesium chloride, 1.3M solution in THF, AcroSeal™, Thermo Scientific™

Product Code. 10361280 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10361280 100 mL AcroSeal™ Glass Bottle 100mL
10179489 800 mL AcroSeal™ Glass Bottle 800mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10361280 Supplier Thermo Scientific Acros Supplier No. 436211000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6393-56-2 , 109-99-9
Molecular Formula C5H11ClMg
Molecular Weight (g/mol) 130.90
MDL Number MFCD00672089
InChI Key OHLJPYMGJFINNA-UHFFFAOYSA-M
Synonym pentylmagnesium chloride, pentylmagnesium chloride solution, n-pentylmagnesium chloride, 1-pentylmagnesium chloride, chloro pentyl magnesium, ohljpymgjfinna-uhfffaoysa-m, 1-pentylmagnesium chloride, 1m in methf, pentylmagnesium chloride solution, 2.0 m in thf
PubChem CID 10953547
IUPAC Name magnesium;pentane;chloride
SMILES CCCCC[Mg]Cl

Specifications

Density 0.9670g/mL
Boiling Point 65.0°C
Flash Point −17°C
Packaging AcroSeal™ Glass Bottle
Quantity 100 mL
Specific Gravity 0.967
Solubility Information Solubility in water: reacts.
Formula Weight 130.9
Physical Form Liquid
Chemical Name or Material Pentylmagnesium chloride
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Pentylmagnesium chloride, 2M solution in THF
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
  • Substance which, in contact with water, emits flammable gases Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.