missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ PEP 005

Product Code. 30001471
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001471 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30001471 Supplier R&D Systems™ Supplier No. 4054/1
  • ONLINE EXCLUSIVE PRICE
    339.67 €
    304.50 € / 1mg
    Save 35.17 €
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Protein kinase C activator

Activator of protein kinase C (PKC). Exhibits antileukemic activity; induces nuclear translocation of PKCδ, and induces apoptosis in acute myeloid leukemia (AML) cell lines and primary AML blast cells. Also displays antiproliferative effects and induces apoptosis in Colo205 cells. Reactivates latent HIV in cell culture. Also acts as a substrate for multidrug transporter.
TRUSTED_SUSTAINABILITY

Specifications

CAS 75567-37-2
Quantity 1 mg
Product Line Tocris
Molecular Formula C25H34O6
Formula Weight 430.53
Purity >98%
Molecular Weight (g/mol) 430.54
Chemical Name or Material PEP 005
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.