missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ PepMap™ 300 C4 HPLC Columns

Use this nanocolumn, packed with 300Å wide pore silica for high-resolution analyses of hydrophobic proteins and peptides.

Brand:  Thermo Scientific™ 164721

Code : Z1

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 13401218

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

Separate hydrophobic peptides and proteins, providing higher recoveries with Thermo Scientific™ Acclaim™ PepMap™ 300 C4 LC Columns.

  • High resolution in protein identification, biomarker discovery and systems biology
  • Particular loadability allows hghest sensitivity in LC/MS
  • Designed for TFA-free LC-MS, minimizing ion-suppression effects
  • Optimized for coupling to ESI/MS and MALDI-MS
  • Highest column-to-column reproducibility
  • Easy-to-use, cutting-edge miniaturized HPLC
Specifications

Specifications

Analytical Column, NanoViper
LC/MS, ESI-MS, MALDI-MS
7250 psi (500 bar)
2 to 8
450 m²/g
L26
nanoLC
3%
Yes
Analytical Column, NanoViper
1 mm
150 mm
5 μm
300 Å
60°C
C4
Silica
Reversed Phase
Reversed Phase
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.