missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Pepsin, for biochemistry, powder
Shop All Thermo Scientific Products
Click to view available options
Quantity:
20 g
100 g
500 g
2.5 kg
Unit Size:
100g
2.5kg
20g
500g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
- Pepsin is an endopeptidase that digests proteins by hydrolyzing peptide bonds
- It is the active form of pepsinogen, which is produced in the chief cells of the gastric mucosa
Applications
- Pepsin is commonly used to produce F(ab’)2 fragments from antibodies
- It has been used along with other enzymes to demonstrate the effects of fixation and enzymatic digestion in immunohistochemical assays, using paraffin-embedded tissue
- It can be used in hydrolyzing dry cervical samples in mice
- It has been used in the digestion of crude wheat gliadin
- It has been used to stimulate in vitro gastric digestion of cooked cod, cocoa mass, and dietary fiber
Specifications
Specifications
| Quantity | 20 g |
| Product Type | Pepsin |
Safety and Handling
Product Identifier
- Pepsin
- Danger
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : Pepsin A
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction