missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Pepsin from Porcine Stomach

Product Code. 15875178 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
25 g
1 kg
Unit Size:
1kg
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15875178 25 g 25g
15845328 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15875178 Supplier Thermo Scientific Chemicals Supplier No. J20010.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Derived from Porcine stomach

  • One unit causes an increase in absorbance at 280nm of 0.001/min. at 37°C at pH 2.0.
TRUSTED_SUSTAINABILITY

Tekniske data

CAS 9001-75-6
Form Lyophilized powder
Quantity 25 g
Synonym EC 3.4.23.1,Pepsin A
Product Type Pepsin

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.