missing translation for 'onlineSavingsMsg'
Learn More

Melford Pepstatin A

Product Code. 30018677
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018677 5 mg 5mg
30018678 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30018677 Supplier Melford Supplier No. P301000.005
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

An inhibitor of aspartic proteases.

An inhibitor of aspartic proteases. Forms a 1:1 complex with proteases such as pepsin, renin and cathepsin D. Highly selective and does not inhibit thiol proteases, neutral proteases, or serine proteases.

Specifications

CAS 26305-03-3
Molecular Formula C34H63N5O9
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.