Learn More
Thermo Scientific Chemicals Pepstatin A, 98%
Description
Pepstatin A, is used as an irreversible inhibitor of HIV-1, aspartic proteases. Inhibits lysosomal proteases and interferes with autolysosomal digestion when used in combination with E 64d. Pepstatin A also efficiently inhibits HIV-1 protease and Fcγ receptor induction by IFN-γ.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Tekniske data
Tekniske data
| For Use With (Application) | Protease inhibitor that acts on pepsin, renin and cathepsin D |
| Molecular Weight (g/mol) | 685.90 |
| Purity | 98% |
| Molecular Formula | C34H63N5O9 |
| Content And Storage | 2°C to 8°C |
| Beilstein | 2201362 |
| Form | Powder |
| InChI Key | FAXGPCHRFPCXOO-JHTFMCFMSA-N |
| SMILES | CC(C)CC(NC(=O)[C@H](C)NC(=O)CC(O)C(CC(C)C)NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)CC(C)C)C(C)C)C(C)C)C(O)CC(O)=O |
| IUPAC Name | 3-hydroxy-4-[(2S)-2-{3-hydroxy-6-methyl-4-[(2S)-3-methyl-2-[(2S)-3-methyl-2-(3-methylbutanamido)butanamido]butanamido]heptanamido}propanamido]-6-methylheptanoic acid |
| Vis mere |
RUO – Research Use Only
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.