missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Pepstatin A, 98%

Product Code. 15483039
Change view
Click to view available options
Quantity:
5 mg
25 mg
Product Type:
Pepstatin
Protease Inhibitor
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Product Type unitSize
15483039 25 mg Protease Inhibitor 25mg
15473039 5 mg Pepstatin 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15483039 Supplier Thermo Scientific Chemicals Supplier No. J60237.MB
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Protease inhibitor that acts on pepsin, renin and cathepsin D | CAS: 26305-03-3 | C34H63N5O9, C34H63N5O9 | 685.90 g/mol

Pepstatin A, is used as an irreversible inhibitor of HIV-1, aspartic proteases. Inhibits lysosomal proteases and interferes with autolysosomal digestion when used in combination with E 64d. Pepstatin A also efficiently inhibits HIV-1 protease and Fcγ receptor induction by IFN-γ.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26305-03-3

Specifications

For Use With (Application) Protease inhibitor that acts on pepsin, renin and cathepsin D
Molecular Weight (g/mol) 685.90g/mol, 685.90
Purity 98%
Molecular Formula C34H63N5O9, C34H63N5O9
Content And Storage 2°C to 8°C
Beilstein 2201362
Form Powder
InChI Key FAXGPCHRFPCXOO-JHTFMCFMSA-N
SMILES CC(C)CC(NC(=O)[C@H](C)NC(=O)CC(O)C(CC(C)C)NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)CC(C)C)C(C)C)C(C)C)C(O)CC(O)=O
IUPAC Name 3-hydroxy-4-[(2S)-2-{3-hydroxy-6-methyl-4-[(2S)-3-methyl-2-[(2S)-3-methyl-2-(3-methylbutanamido)butanamido]butanamido]heptanamido}propanamido]-6-methylheptanoic acid
Quantity 25 mg
Product Type Protease Inhibitor
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.