missing translation for 'onlineSavingsMsg'
Learn More

Pepstatin A, Fisher BioReagents

Product Code. 10786834
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
100 mg
250 mg
Packaging:
Amber Glass
Unit Size:
100mg
10mg
250mg
25mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10786834 10 mg Amber Glass 10mg
10011413 5 mg Amber Glass 5mg
10264583 25 mg Amber Glass 25mg
10410464 100 mg Amber Glass 100mg
10051553 250 mg Amber Glass 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10786834 Supplier Fisher BioReagents Supplier No. BP267110
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Pepstatin A is an inhibitor for pepsin, renin, cathepsin D, and other acid proteases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26305-03-3
Molecular Formula C34H63N5O9
Molecular Weight (g/mol) 685.904
InChI Key FAXGPCHRFPCXOO-WQILDUGRSA-N
Synonym pepstatin a
PubChem CID 131801262
IUPAC Name (3R,4S)-3-hydroxy-4-[[(2R)-2-[[(3R,4S)-3-hydroxy-6-methyl-4-[[(2S)-3-methyl-2-[[(2S)-3-methyl-2-(3-methylbutanoylamino)butanoyl]amino]butanoyl]amino]heptanoyl]amino]propanoyl]amino]-6-methylheptanoic acid
SMILES CC(C)CC(C(CC(=O)O)O)NC(=O)C(C)NC(=O)CC(C(CC(C)C)NC(=O)C(C(C)C)NC(=O)C(C(C)C)NC(=O)CC(C)C)O

Specifications

Melting Point 235°C
Color White
Melting Range 230° to 240°C
Infrared Spectrum Conforms to reference
Packaging Amber Glass
Quantity 10 mg
Merck Index 15, 7260
Formula Weight 685.9
Physical Form Powder or Solid
Chemical Name or Material Pepstatin A
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.