missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Peracetic Acid 2000 Test Strip

Product Code. 11817073 Shop All Macherey-Nagel Products
Click to view available options
Quantity:
100 Pack
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 11817073

Brand: Macherey-Nagel™ 91342

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Graduations: 0 to 2000mg/L

Order Info

Graduations

Specifications

Format Semi Quantitative Test Strips
For Use With (Application) For the fast and reliable determination of peracetic acid in solutions
Graduations 0 to 2000mg/L
Quantity 100 Pack
Type Peracetic Acid 2000 Test Strip
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.