missing translation for 'onlineSavingsMsg'
Learn More

Peracetic Acid Solution, 0%, Honeywell™ Fluka™

Product Code. 15672820 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
5 mL
100 mL
500 mL
Unit Size:
100mL
500mL
5mL
This item is not returnable. View return policy

Product Code. 15672820

Brand: Honeywell Research Chemicals 269336500ML

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 79-21-0
Molecular Formula C2H4O3
Molecular Weight (g/mol) 76.051
MDL Number MFCD00002128
InChI Key KFSLWBXXFJQRDL-UHFFFAOYSA-N
Synonym peracetic acid, peroxyacetic acid, estosteril, acetic peroxide, peroxoacetic acid, monoperacetic acid, osbon ac, acetyl hydroperoxide, proxitane 4002, desoxon 1
PubChem CID 6585
IUPAC Name ethaneperoxoic acid
SMILES CC(=O)OO

Specifications

Density 1.13g/mL at 25°C
Linear Formula CH3CO3H
Quantity 500 mL
Beilstein 1098464
Formula Weight 76.05g/mol
Vapor Pressure 20mmHg (25°C)
Percent Purity 0%
Chemical Name or Material Peracetic Acid Solution
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.