missing translation for 'onlineSavingsMsg'
Learn More

Peracetic Acid Solution, purum, Honeywell™

Product Code. 15627060 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
500mL
This item is not returnable. View return policy

Product Code. 15627060

Brand: Honeywell-Fluka 77240100ML

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 79-21-0
Molecular Formula C2H4O3
Molecular Weight (g/mol) 76.051
MDL Number MFCD00002128
InChI Key KFSLWBXXFJQRDL-UHFFFAOYSA-N
Synonym peracetic acid, peroxyacetic acid, estosteril, acetic peroxide, peroxoacetic acid, monoperacetic acid, osbon ac, acetyl hydroperoxide, proxitane 4002, desoxon 1
PubChem CID 6585
IUPAC Name ethaneperoxoic acid
SMILES CC(=O)OO

Specifications

Density 1.15g/mL at 20°C
Packaging Glass bottle
Linear Formula CH3CO3H
Quantity 100 mL
Beilstein 1098464
Formula Weight 76.05g/mol
Grade purum p.a.
Concentration or Composition (by Analyte or Components) 39% in acetic acid
Chemical Name or Material Peracetic acid solution
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Peracetic acid, ca 39% in acetic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Organic peroxide Type C & D
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3
Hazard Statement
  • H242-Heating may cause a fire.
  • H301-Toxic if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Acetic acid, Peroxyacetic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.