missing translation for 'onlineSavingsMsg'
Learn More

Perchloric Acid, 0.7, ULTREX™ II Ultrapure Reagent, J.T. Baker™

Product Code. 15528314 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
PFA Bottle
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15528314 500 mL PFA Bottle Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15528314 Supplier Avantor J.T.Baker Supplier No. 480601
  • ONLINE EXCLUSIVE PRICE
    1368.53 €
    1209.65 € / Pack of 1
    Save 158.88 €
    11% Off
 Restricted Item
Estimated Shipment: 05-03-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reagents for critical elemental analysis with less than 10 parts-per-trillion (ppt) levels of up to 65 elements

    For use in ICP (Inducutively Coupled Plasma Spectrometry), Trace Metal Analysis; For Laboratory,Research,or Manufacturing Use

Specifications

CAS 7601-90-3
Packaging PFA Bottle
Specific Gravity 1L = 1.60kg
Quantity 500 mL
Molecular Formula ClHO4
MDL Number MFCD00011325
Synonym unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4
InChI Key VLTRZXGMWDSKGL-UHFFFAOYSA-N
SMILES O[Cl](=O)(=O)=O
IUPAC Name perchloric acid
Molecular Weight (g/mol) 100.45
PubChem CID 24247
ChEBI CHEBI:29221
Formula Weight 100.46
Percent Purity 70%
Grade ULTREX™ II Ultrapure Reagent
Chemical Name or Material Perchloric Acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.