missing translation for 'onlineSavingsMsg'
Learn More

Perchloric Acid 70% d=1.67, Certified AR for Analysis, Fisher Chemical™

Product Code. 10234490 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Unit Size:
1 L
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10234490 500 mL 500mL
10538820 1 L 1 L
10548820 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10234490 Supplier Fisher Chemical Supplier No. P/1280/PB08
  • ONLINE EXCLUSIVE PRICE
    162.68 €
    107.00 € / 500mL
    Save 55.68 €
    34% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 7601-90-3
Molecular Formula ClHO4
Molecular Weight (g/mol) 100.45
MDL Number MFCD00011325
InChI Key VLTRZXGMWDSKGL-UHFFFAOYSA-N
Synonym unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4
PubChem CID 24247
ChEBI CHEBI:29221
IUPAC Name perchloric acid
SMILES O[Cl](=O)(=O)=O

Specifications

pH 0.1
Color Colorless
Melting Point -18°C
Boiling Point 203°C
Packaging Plastic Coated Glass bottle
Vapor Pressure 6.8mmHg 25
Quantity 500 mL
Viscosity 3.5 mPaS at 20°C
Physical Form Liquid
Formula Weight 100.46g/mol
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Perchloric acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Oxidising liquid Category 1
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H373-May cause damage to organs.
  • H302-Harmful if swallowed.
  • H290-May be corrosive to metals.
  • H271-May cause fire or explosion; strong oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.