missing translation for 'onlineSavingsMsg'
Learn More

Perchloric Acid Solution, Solstice

Product Code. 15644170
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass bottle
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15644170 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15644170 Supplier Honeywell-Fluka Supplier No. 344081L
  • ONLINE EXCLUSIVE PRICE
    507.00 €
    457.52 € / 1 L
    Save 49.48 €
    9% Off
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Volumetric, 0.01 M HClO4 in acetic acid (0.01N), for titration in non-aqueous liquids

Solstice's high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use
  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations
TRUSTED_SUSTAINABILITY

Specifications

pH Acidic
Color Colorless
CAS 7601-90-3
Boiling Point 119°C (1013hPa)
Density ∽1.06g/cu.cm at 20°C
Odor Stinging
Packaging Glass bottle
Flash Point 40°C (104°F)
Quantity 1 L
Molecular Formula ClHO4
Linear Formula HClO4
Physical Form Liquid
MDL Number MFCD00011325
UN Number UN2920
Synonym unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4
Solubility Information Completely miscible in water
InChI Key VLTRZXGMWDSKGL-UHFFFAOYSA-N
SMILES O[Cl](=O)(=O)=O
IUPAC Name perchloric acid
Molecular Weight (g/mol) 100.45
PubChem CID 24247
ChEBI CHEBI:29221
Formula Weight 100.46g/mol
Chemical Name or Material Perchloric acid solution, Volumetric, 0.01 M HClO4 in acetic acid (0.01N), for titration in non-aqueous liquids
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.