missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Percoll™ PLUS Centrifugation Media

Product Code. 10166144 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
250 mL
1 L
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10166144 1 L 1 L
11500744 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10166144 Supplier Cytiva Supplier No. 17544501
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Low endotoxin reagent for density gradient centrifugation of cells, viruses, and subcellular particles. Cytiva™ Percoll™ PLUS Centrifugation Media is non-toxic, shows low viscosity and is ideal for density gradients.

  • Gradients can either be preformed or spontaneously generated by centrifugation at moderate speeds in an angle-head rotor
  • Low viscosity of media allows cell isolations on preformed gradients to be completed in only a few minutes using low centrifugal forces (200 to 1000xG)
  • Iso-osmotic gradients throughout cover a range of densities up to 1.3g/mL
  • Nontoxic, almost chemically inert and does not adhere to membranes
  • Adjustable to physiological ionic strength and pH; can be buffered within pH range 5.5 to 10.0 without change in properties
  • Silica sol coated with covalently linked silane; 15-30nm dia. bead size
  • Low endotoxin levels (max. 2 EU/mL); well-suited for cell separation in clinical research applications

For in vitro research use only.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 L
Content And Storage 4–30°C
For Use With (Application) Cell Isolation
Viscosity 15cP
Endotoxin Level 2 EU/mL
Osmolality < 30 mOsm/kg
Color Colorless to Slightly Yellow
pH 8.9 to 9.9
Product Type Centrifugation Media
Density 1.135 g/mL
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.