missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ PERFECT CUTTING™ Diamond Gloves

Product Code. 15826775
Change view
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
1 pair
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15826775 8 1 pair
15806775 6 1 pair
15816775 7 1 pair
15836775 9 1 pair
15846775 10 1 pair
15856775 11 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 15826775 Supplier Honeywell™ Supplier No. 233224508
  • ONLINE EXCLUSIVE PRICE
    31.31 €
    13.86 € / 1 pairs
    Save 17.45 €
    55% Off
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Honeywell™ PERFECT CUTTING™ Diamond Gloves for handling sharp parts in a dry or greasy environment.

  • White Dyneema™ Diamond/lycra™ knitted glove
  • Black polyurethane coating on palm and tips of fingers
  • The knitting technique helps to obtain a glove that conforms snugly to the hand and good dexterity
  • The glove offers maximum comfort and added sturdiness
  • The cool Dyneema™ diamond touch gives an added sensation of comfort
  • The tight knit (gauge 13) gives the glove perfect support, limits the penetration of dirt and offers a good touch
  • The glove's ventilated back optimises its breathability
  • The Dyneema™ diamond/lycra™ thread provides outstanding properties against cuts
  • The polyurethane coating at the palm and fingertips provides outstanding abrasion resistance
  • Its white color enhances the glove's hygiene and cleanliness
  • Its black coating enables work in a slightly soiled environment
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EN 420-2003 EEC Basic standards, EN 388-2003 Mechanical hazards
Description 13 gauge
Color White, Gray
Material Polyurethane, Lycra
Size 8
Length (Metric) 23.5 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.