Learn More
Honeywell™ PERFECT POLY™ PU First Gray Gloves
Product Code. 15826685
Click to view available options
Size:
6
7
8
9
Unit Size:
1 pair
Each
Product Code. 15826685
Brand: Honeywell™ 210025006
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Honeywell™ PERFECT POLY™ PU First Gray Gloves for handling of fine objects in a dry environment, or slightly greasy and dirty objects.
  • Gray polyamide knitted glove
  • Gray polyurethane coating at the palm and fingertips
  • The knitting technique helps to obtain a glove that conforms snugly to the hand, with good dexterity
  • The glove offers maximum comfort and added sturdiness
  • The tight knit gives the glove perfect support, limits the penetration of dirt and offers a good touch
  • The ventilated back and PU coating optimise the glove's breathability
  • Polyamide with its polyurethane coating provide excellent abrasion and tear resistance
  • The PU coating ensures a good grip
  • Non pilling, the polyamide is suitable for precision work
  • The gloves are certified as silicone-free to ensure their use in painting environment
  • Gray color makes it more soilproof, thus ensuring a longer life
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 420-2003 Basic standard, EN 388-2016 Mechanical risk
Description Knitted Coating, 13 gauge
Coating Material Polyurethane
Color Gray
Material Polyamide
Size 6
Length (Metric) 25 cm