Learn More
Honeywell™ PERFECT POLY™ Skin Gloves
Product Code. 15836795
Change view
Click to view available options
Size:
7
8
9
Unit Size:
1 pair
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15836795 7 1 pair
15846795 8 1 pair
15856795 9 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15836795 Supplier Honeywell™ Supplier No. 240026007
Add to basket
Añadir a la cesta
Este artículo no se puede devolver. Vea la política de devoluciones
Honeywell™ PERFECT POLY™ Skin Gloves for handling fine products in a dry or slightly greasy or soiled environment.
  • Polyamide knitted glove
  • Gray polyurethane coating on palm and tips of fingers
  • Elastic wrist
  • The knitting technique helps to obtain a glove that conforms snugly to the hand as well as outstanding dexterity
  • The glove offers maximum comfort and added sturdiness
  • The ultra tight weave gives the glove perfect support, limits the penetration of dirt and offers a good touch
  • The ventilated back and ultra light coating optimise the glove's breathability
  • Polyamide with its polyurethane coating provide excellent abrasion and tear resistance
  • Non pilling, the polyamide is suitable for precision work
  • The gloves are certified as silicone-free to ensure their use in painting
  • Blue color and gray coating make the glove more soilproof, thus ensuring a longer life
TRUSTED_SUSTAINABILITY
Especificaciones
Certifications/Compliance EN 420-2003 Basic standads, EN 388-2016 Mechanical hazards
Description 18 gauge
Color Blue, Gray
Material Polyamide, Polyurethane
Size 7
Length (Metric) 24 cm
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.