missing translation for 'onlineSavingsMsg'
Learn More

Perfluoro-1,3,5-trimethylcyclohexane, mixture of isomers, tech., Thermo Scientific Chemicals

Product Code. 11497044
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11497044 50 g 50g
11487044 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11497044 Supplier Thermo Scientific Alfa Aesar Supplier No. B21500.18
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

It finds its application in the characterization of non-aqueous phase liquid/tracer interaction in support of a vadose zone partitioning interwell tracer test.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in the characterization of non-aqueous phase liquid/tracer interaction in support of a vadose zone partitioning interwell tracer test.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 374-76-5
Molecular Formula C9F18
Molecular Weight (g/mol) 450.07
MDL Number MFCD00066614
InChI Key MGOFOLPXKULBGG-UHFFFAOYSA-N
Synonym perfluoro-1,3,5-trimethylcyclohexane, 1,1,2,3,3,4,5,5,6-nonafluoro-2,4,6-tris trifluoromethyl cyclohexane, acmc-1ajc4, nonafluoro-1,3,5-tris-trifluoromethyl-cyclohexane, 1,2,2,3,4,4,5,6,6-nonafluoro-1,3,5-tris trifluoromethyl cyclohexane, cyclohexane,1,1,2,3,3,4,5,5,6-nonafluoro-2,4,6-tris trifluoromethyl, 1,1,2,3,3,4,5,5,6-nonakis fluoranyl-2,4,6-tris trifluoromethyl cyclohexane
PubChem CID 2776392
IUPAC Name 1,1,2,3,3,4,5,5,6-nonafluoro-2,4,6-tris(trifluoromethyl)cyclohexane
SMILES C1(C(C(C(C(C1(F)F)(C(F)(F)F)F)(F)F)(C(F)(F)F)F)(F)F)(C(F)(F)F)F

Specifications

Melting Point -68°C
Density 1.888
Boiling Point 125°C to 128°C
Refractive Index 1.297
Quantity 50 g
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 450.07
Grade Technical
Chemical Name or Material Perfluoro-1,3,5-trimethylcyclohexane, mixture of isomers

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.