missing translation for 'onlineSavingsMsg'
Learn More

Perfluoro-1-iododecane, TRC

Product Code. 17914975
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17914975 5 g 5g
17902465 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17914975 Supplier Toronto Research Chemicals Supplier No. TRCP2865305G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 423-62-1
Molecular Formula C10 F21 I
Molecular Weight (g/mol) 645.978
Synonym Heneicosafluoro-1-iodo-decane, 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10-heneicosafluoro-10-iododecane, 1-Iodoheneicosafluorodecane, 1-Iodoperfluorodecane, Heneicosafluoro-1-iododecane, Perfluorodecyl iodide
IUPAC Name 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10-henicosafluoro-10-iododecane
SMILES FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)I

Specifications

Quantity 5 g
InChI Formula InChI=1S/C10F21I/c11-1(12,3(15,16)5(19,20)7(23,24)9(27,28)29)2(13,14)4(17,18)6(21,22)8(25,26)10(30,31)32
Formula Weight 645.871
Percent Purity >95%
Chemical Name or Material Perfluoro-1-iododecane
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.