missing translation for 'onlineSavingsMsg'
Learn More

Perfluoro-1-iodooctane, 98%

Product Code. 11429656
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11429656 10 g 10g
11439656 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11429656 Supplier Thermo Scientific Alfa Aesar Supplier No. B20207.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 507-63-1 | C8F17I | 545.965 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 507-63-1
Molecular Formula C8F17I
Molecular Weight (g/mol) 545.965
MDL Number MFCD00001064
InChI Key KWXGJTSJUKTDQU-UHFFFAOYSA-N
Synonym perfluorooctyl iodide, heptadecafluoro-1-iodooctane, 1-iodoperfluorooctane, perfluoro-n-octyl iodide, 1-iodoheptadecafluorooctane, perfluoro-1-iodooctane, perfluorooctyliodide, heptadecafluoro-n-octyl iodide, octane, 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-heptadecafluoro-8-iodo, octane, heptadecafluoro-1-iodo
PubChem CID 10491
IUPAC Name 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-heptadecafluoro-8-iodooctane
SMILES C(C(C(C(C(F)(F)I)(F)F)(F)F)(F)F)(C(C(C(F)(F)F)(F)F)(F)F)(F)F

Specifications

Melting Point 25°C
Density 2.04
Boiling Point 160°C to 161°C
Linear Formula CF3(CF2)7I
Refractive Index 1.329
Quantity 10 g
Beilstein 1717141
Sensitivity Light sensitive
Formula Weight 545.97
Percent Purity 98%
Chemical Name or Material Perfluoro-1-iodooctane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.