missing translation for 'onlineSavingsMsg'
Learn More

Perfluoro(4-methyl-2-pentene), [(E):(Z) 9:1], 90+%, cont 5-10% perfluoro(2-methyl-2-pentene)

Product Code. 11313786
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11313786 25 g 25g
11323786 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11313786 Supplier Thermo Scientific Alfa Aesar Supplier No. L16836.14
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 2070-70-4 | C6F12 | 300.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2070-70-4
Molecular Formula C6F12
Molecular Weight (g/mol) 300.05
MDL Number MFCD00153253
InChI Key SAPOZTRFWJZUFT-UHFFFAOYSA-N
Synonym 2z-1,1,1,2,3,4,5,5,5-nonafluoro-4-trifluoromethyl pent-2-ene
PubChem CID 11012007
SMILES FC(=C(F)C(F)(C(F)(F)F)C(F)(F)F)C(F)(F)F

Specifications

Linear Formula (CF3)2CFCF=CFCF3
Refractive Index <1.3000
Quantity 25 g
IUPAC Name 1,1,1,2,3,4,5,5,5-nonafluoro-4-(trifluoromethyl)pent-2-ene
Formula Weight 300.05
Percent Purity ≥90%
Assay contains 5 to 10% perfluoro(2-methyl-2-pentene)
Chemical Name or Material Perfluoro(4-methyl-2-pentene), [(E):(Z) 9:1]

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.