missing translation for 'onlineSavingsMsg'
Learn More

Perfluoro(4-methyl-2-pentenoic acid), 97%, Thermo Scientific™

Product Code. 11429327
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
This item is not returnable. View return policy

Product Code. 11429327

Brand: Thermo Scientific Alfa Aesar B21376.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103229-89-6
Molecular Formula C6HF9O2
Molecular Weight (g/mol) 276.06
MDL Number MFCD00153286
InChI Key TWDHAMRDYDLSPH-UHFFFAOYSA-N
Synonym z-2,3,4,5,5,5-hexafluoro-4-trifluoromethyl pent-2-enoic acid, 2z-2,3,4,5,5,5-hexafluoro-4-trifluoromethyl pent-2-enoic acid
PubChem CID 44717505
SMILES OC(=O)C(F)=C(F)C(F)(C(F)(F)F)C(F)(F)F

Specifications

Boiling Point 173°C to 174°C
Quantity 10 g
UN Number UN3265
IUPAC Name (Z)-2,3,4,5,5,5-hexafluoro-4-(trifluoromethyl)pent-2-enoic acid
Formula Weight 276.06
Percent Purity 97%
Chemical Name or Material Perfluoro(4-methyl-2-pentenoic acid)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.