missing translation for 'onlineSavingsMsg'
Learn More

Perfluoro(butylcyclohexane), TRC

Product Code. 30418664
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30418664 500 mg 500mg
30413493 1 g -
30415974 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30418664 Supplier Toronto Research Chemicals Supplier No. TRCP999370500MG
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 374-60-7
Molecular Formula C10F20
Molecular Weight (g/mol) 500.08
Synonym 1,1,2,2,3,3,4,4,5,5,6-undecafluoro-6-(1,1,2,2,3,3,4,4,4-nonafluorobutyl)-Cyclohexane, undecafluoro(nonafluorobutyl)-Cyclohexane, 1,1,2,2,3,3,4,4,5,5,6-Undecafluoro-6-(1,1,2,2,3,3,4,4,4-nonafluorobutyl)cyclohexane
IUPAC Name 1,1,2,2,3,3,4,4,5,5,6-undecafluoro-6-(1,1,2,2,3,3,4,4,4-nonafluorobutyl)cyclohexane
SMILES FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C1(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C1(F)F

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C10F20/c11-1(4(16,17)7(22,23)9(26,27)10(28,29)30)2(12,13)5(18,19)8(24,25)6(20,21)3(1,14)15
Formula Weight 499.9681
Chemical Name or Material Perfluoro(butylcyclohexane)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.