missing translation for 'onlineSavingsMsg'
Learn More

Perfluoro-compound FC-40(TM), Thermo Scientific Chemicals

Product Code. 10163360 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
100 g
250 g
Packaging:
Glass bottle
Unit Size:
100g
10g
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10163360 10 g Glass bottle 10g
10081150 100 g Glass bottle 100g
10173360 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10163360 Supplier Thermo Scientific Acros Supplier No. 123760100
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51142-49-5
Molecular Formula C21F48N2
Molecular Weight (g/mol) 1192.168
MDL Number MFCD01632188
InChI Key QDOIZVITZUBGOQ-UHFFFAOYSA-N
Synonym fluorinert fc-40, perfluoro-compound fc-40, bis nonafluorobutyl trifluoromethyl amine; perfluorotributylamine, 1,1,2,2,3,3,4,4,4-nonafluoro-n,n-bis 1,1,2,2,3,3,4,4,4-nonafluorobutyl butan-1-amine,1,1,2,2,3,3,4,4,4-nonafluoro-n-1,1,2,2,3,3,4,4,4-nonafluorobutyl-n-trifluoromethyl butan-1-amine, 1,1,2,2,3,3,4,4,4-nonafluoro-n,n-bis 1,1,2,2,3,3,4,4,4-nonafluorobutyl butan-1-amine; 1,1,2,2,3,3,4,4,4-nonafluoro-n-1,1,2,2,3,3,4,4,4-nonafluorobutyl-n-trifluoromethyl butan-1-amine, bis 1,1,2,2,3,3,4,4,4-nonafluorobutyl trifluoromethyl amine; perfluorotributylamine
PubChem CID 2723673
IUPAC Name 1,1,2,2,3,3,4,4,4-nonafluoro-N,N-bis(1,1,2,2,3,3,4,4,4-nonafluorobutyl)butan-1-amine;1,1,2,2,3,3,4,4,4-nonafluoro-N-(1,1,2,2,3,3,4,4,4-nonafluorobutyl)-N-(trifluoromethyl)butan-1-amine
SMILES C(C(C(F)(F)F)(F)F)(C(N(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F.C(C(C(F)(F)F)(F)F)(C(N(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)C(F)(F)F)(F)F)(F)F

Specifications

Color Colorless
Density 1.8700g/mL
Boiling Point 158.0°C to 173.0°C
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.29
Quantity 10 g
Specific Gravity 1.87
Viscosity 1.6-2.8 mm2/s (25°C)
Formula Weight 650
Physical Form Liquid
Chemical Name or Material Perfluoro-compound FC-40™
Show More Show Less
Product Identifier
  • Perfluoro-compound FC-40|TS
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 4
Hazard Statement
  • H413-May cause long lasting harmful effects to aquatic life.
Precautionary Statement
  • P273-Avoid release to the environment.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.