missing translation for 'onlineSavingsMsg'
Learn More

Perfluoro(decahydronaphthalene), cis + trans, 95%, Thermo Scientific Chemicals

Product Code. 11433837
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11433837 500 g 500g
11413837 25 g 25g
11423837 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex I to EU Regulation No. 517/2014 on fluorinated greenhouse gases, all customers must complete a declaration before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11433837 Supplier Thermo Scientific Alfa Aesar Supplier No. A18288.36
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex I to EU Regulation No. 517/2014 on fluorinated greenhouse gases, all customers must complete a declaration before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Perfluoro(decahydronaphthalene) is used to dissolve gases. Due to its gas carrying capacity, it is utilized to increase oxygen delivery during cell culture. It acts as an ingredient in fluosol which is an artificial blood product. It is also used to dissolve Teflon.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Perfluoro(decahydronaphthalene) is used to dissolve gases. Due to its gas carrying capacity, it is utilized to increase oxygen delivery during cell culture. It acts as an ingredient in fluosol which is an artificial blood product. It is also used to dissolve Teflon.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 306-94-5
Molecular Formula C10F18
Molecular Weight (g/mol) 462.081
MDL Number MFCD00010626
InChI Key UWEYRJFJVCLAGH-UHFFFAOYSA-N
Synonym perfluorodecalin, perflunafene, octadecafluorodecahydronaphthalene, cis-perfluorodecalin, trans-perfluorodecalin, perfluorodecahydronaphthalene, naphthalene, octadecafluorodecahydro, f-dc, pp 5, perflunafenum latin
PubChem CID 9386
ChEBI CHEBI:38848
IUPAC Name 1,1,2,2,3,3,4,4,4a,5,5,6,6,7,7,8,8,8a-octadecafluoronaphthalene
SMILES C12(C(C(C(C(C1(F)F)(F)F)(F)F)(F)F)(C(C(C(C2(F)F)(F)F)(F)F)(F)F)F)F

Specifications

Melting Point -10°C to -7°C
Density 1.94
Boiling Point 141°C to 142°C
Refractive Index 1.3145
Quantity 500 g
Beilstein 2067113
Merck Index 14,4183
Solubility Information Insoluble in water.
Formula Weight 462.1
Percent Purity 95%
Chemical Name or Material Perfluoro(decahydronaphthalene), cis + trans
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.