missing translation for 'onlineSavingsMsg'
Learn More

Perfluorobutanoic acid 13C3 (2,3,4-13C3) 50 μg/mL in Methanol:Water, Dr. Ehrenstorfer

Product Code. 30325202
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30325202 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30325202 Supplier Dr. Ehrenstorfer Supplier No. DREA15986523MW50
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Perfluorobutanoic acid 13C3 (2,3,4-13C3)
CAS 2483735-33-5
Solvent or Matrix Methanol/Water
Molecular Formula 13C3 C H F7 O2
Quantity 1 mL
Synonym 13C3-PFBA, Perfluorobutanoic Acid 13C3 (2,3,4-13C3), Butanoic-2,3,4-13C3 acid, 2,2,3,3,4,4,4-heptafluoro- (ACI)
InChI Formula InChI=1S/C4HF7O2/c5-2(6,1(12)13)3(7,8)4(9,10)11/h(H,12,13)/i2+1,3+1,4+1
SMILES OC(=O)[13C](F)(F)[13C](F)(F)[13C](F)(F)F
IUPAC Name 2,2,3,3,4,4,4-heptafluoro(2,3,4-^{13}C_{3})butanoic acid
Molecular Weight (g/mol) 217.01633
Formula Weight 216.9965 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.