missing translation for 'onlineSavingsMsg'
Learn More

Perfluorodecanoic Acid, TRC

Product Code. 18082320
Change view
Click to view available options
Quantity:
100 mg
1 g
25 g
Unit Size:
100mg
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18082320 100 mg 100mg
17979759 1 g 1g
18043940 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 18082320 Supplier Toronto Research Chemicals Supplier No. TRCP286220100MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 335-76-2
Molecular Formula C10 H F19 O2
Molecular Weight (g/mol) 514.08
Synonym Decanoic acid, nonadecafluoro- (6CI,7CI,8CI,9CI), 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Nonadecafluorodecanoic acid, Nonadecafluoro-n-decanoic acid, Nonadecafluorodecanoic acid, PFDA, Perfluoro-1-nonanecarboxylic acid, Perfluoro-n-decanoic acid, Perfluorocapric acid
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-nonadecafluorodecanoic acid
SMILES OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C10HF19O2/c11-2(12,1(30)31)3(13,14)4(15,16)5(17,18)6(19,20)7(21,22)8(23,24)9(25,26)10(27,28)29/h(H,30,31)
Formula Weight 513.9673
Percent Purity >95%
Chemical Name or Material Perfluorodecanoic acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.