missing translation for 'onlineSavingsMsg'
Learn More

Perfluoroheptanes, mixed isomers, 97%, Thermo Scientific Chemicals

Product Code. 11353876
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11353876 100 g 100g
11343876 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11353876 Supplier Thermo Scientific Alfa Aesar Supplier No. L16979.22
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 335-57-9
Molecular Formula C7F16
Molecular Weight (g/mol) 388.051
MDL Number MFCD00040339
InChI Key LGUZHRODIJCVOC-UHFFFAOYSA-N
Synonym hexadecafluoroheptane, perfluoroheptane, perfluoro-n-heptane, heptane, hexadecafluoro, perfluoroheptanes, unii-i23zvd1p1l, i23zvd1p1l, heptane, 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,7-hexadecafluoro, perfluoroheptane s, acmc-20akry
PubChem CID 9553
ChEBI CHEBI:38847
IUPAC Name 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,7-hexadecafluoroheptane
SMILES C(C(C(C(F)(F)F)(F)F)(F)F)(C(C(C(F)(F)F)(F)F)(F)F)(F)F

Specifications

Density 1.72
Boiling Point 80°C to 82°C
Linear Formula CF3(CF2)5CF3
Refractive Index 1.265
Quantity 100 g
Beilstein 1716335
Formula Weight 388.05
Percent Purity 98%
Chemical Name or Material Perfluoroheptanes, mixed isomers

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.