missing translation for 'onlineSavingsMsg'
Learn More

Perfluorooctyl bromide, 99%

Product Code. 10641705 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10641705 10 g Glass bottle 10g
11984501 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10641705 Supplier Thermo Scientific Acros Supplier No. 386790100
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 423-55-2 | C8BrF17 | 498.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 423-55-2
Molecular Formula C8BrF17
Molecular Weight (g/mol) 498.97
MDL Number MFCD00042082
InChI Key WTWWXOGTJWMJHI-UHFFFAOYSA-N
Synonym perflubron, 1-bromoperfluorooctane, perfluorooctyl bromide, 1-bromoheptadecafluorooctane, oxygent, perfluoro-n-octyl bromide, imagent gi, perfluorooctylbromide, liquivent, perfluoroctyl bromide
PubChem CID 9873
ChEBI CHEBI:38803
IUPAC Name 1-bromo-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluorooctane
SMILES FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)Br

Specifications

Melting Point 6.0°C
Color Colorless to Tan
Density 1.9300g/mL
Boiling Point 142.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CF3(CF2)7Br
Refractive Index 1.3050 to 1.3070
Quantity 10 g
Merck Index 15,727
Specific Gravity 1.93
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with organic solvents
Viscosity 2.3 mPa.s (20°C)
Formula Weight 498.95
Percent Purity 99%
Physical Form Viscous Solution
Chemical Name or Material Perfluorooctyl bromide
Show More Show Less
compliance-icons
Product Identifier
  • Perfluorooctyl bromide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.