missing translation for 'onlineSavingsMsg'
Learn More

Perfluorotetradecanoic acid, 96%

Product Code. 11303257
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 11303257

Brand: Thermo Scientific Alfa Aesar L13796.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

CAS: 376-06-7 | C14HF27O2 | 714.117 g/mol

Perfluorotetradecanoic acid is used for GC (Gas Chromatography) and LC (Liquid Chromatography) analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Perfluorotetradecanoic acid is used for GC (Gas Chromatography) and LC (Liquid Chromatography) analysis.

Solubility
It is insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 376-06-7
Molecular Formula C14HF27O2
Molecular Weight (g/mol) 714.117
MDL Number MFCD00153263
InChI Key RUDINRUXCKIXAJ-UHFFFAOYSA-N
Synonym perfluorotetradecanoic acid, perfluoromyristic acid, heptacosafluorotetradecanoic acid, tetradecanoic acid, heptacosafluoro, acmc-20ajxq, perfluorotetradecanoic acid, analytical standard
PubChem CID 67822
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,14-heptacosafluorotetradecanoic acid
SMILES C(=O)(C(C(C(C(C(C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O

Specifications

Melting Point 130°C to 132°C
Boiling Point 192°C (60 mmHg)
Quantity 1 g
UN Number UN3261
Beilstein 1811438
Solubility Information It is insoluble in water.
Formula Weight 714.12
Percent Purity 96%
Chemical Name or Material Perfluorotetradecanoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.