missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perfluorotetradecanoic acid, 96%
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Description
Perfluorotetradecanoic acid is used for GC (Gas Chromatography) and LC (Liquid Chromatography) analysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Perfluorotetradecanoic acid is used for GC (Gas Chromatography) and LC (Liquid Chromatography) analysis.
Solubility
It is insoluble in water.
Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Perfluorotetradecanoic acid is used for GC (Gas Chromatography) and LC (Liquid Chromatography) analysis.
Solubility
It is insoluble in water.
Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Specifications
Specifications
| Melting Point | 130°C to 132°C |
| Boiling Point | 192°C (60 mmHg) |
| Quantity | 5 g |
| UN Number | UN3261 |
| Beilstein | 1811438 |
| Solubility Information | It is insoluble in water. |
| Formula Weight | 714.12 |
| Percent Purity | 96% |
| Chemical Name or Material | Perfluorotetradecanoic acid |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction