missing translation for 'onlineSavingsMsg'
Learn More

Perfluorotri-n-butylamine, tech. 90%, Thermo Scientific Chemicals

Product Code. 11498597
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11498597 25 g 25g
11488597 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex I to EU Regulation No. 517/2014 on fluorinated greenhouse gases, all customers must complete a declaration before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11498597 Supplier Thermo Scientific Alfa Aesar Supplier No. A19712.14
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex I to EU Regulation No. 517/2014 on fluorinated greenhouse gases, all customers must complete a declaration before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Perfluorotri-n-butylamine is used in electronic applications viz. liquid burn-in, testing and vapor phase soldering processes. It acts as an indicator fluid to check the leakage and heat transfer fluid for thermal shock testing. It is an active component in fluosol, which is a substitute for artificial blood and fluorinert coolant liquids. Further, it is used as solvent for computer disc drive lubrication. In addition to this, it is used as a calibration chemical in gas chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Perfluorotri-n-butylamine is used in electronic applications viz. liquid burn-in, testing and vapor phase soldering processes. It acts as an indicator fluid to check the leakage and heat transfer fluid for thermal shock testing. It is an active component in fluosol, which is a substitute for artificial blood and fluorinert coolant liquids. Further, it is used as solvent for computer disc drive lubrication. In addition to this, it is used as a calibration chemical in gas chromatography.

Solubility
Miscible with chloroform, cyclohexane, ethyl ether, ethyl acetate, heptane, carbon tetrachloride and petroleum ether. Immiscible with water, methanol, toluene, isopropyl alcohol, benzene and acetone.

Notes
Store in a cool place. Incompatible with powdered metals and alkali metals.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 311-89-7
Molecular Formula C12F27N
Molecular Weight (g/mol) 671.096
MDL Number MFCD00000436
InChI Key RVZRBWKZFJCCIB-UHFFFAOYSA-N
Synonym perfluorotributylamine, tris perfluorobutyl amine, tri perfluorobutyl amine, tris nonafluorobutyl amine, heptacosafluorotributylamine, fluorinert, fluosol 43, perfluorotri-n-butylamine, fluorocarbon fc 43, tri nonafluorobutyl amine
PubChem CID 9397
ChEBI CHEBI:38854
IUPAC Name 1,1,2,2,3,3,4,4,4-nonafluoro-N,N-bis(1,1,2,2,3,3,4,4,4-nonafluorobutyl)butan-1-amine
SMILES C(C(C(F)(F)F)(F)F)(C(N(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F

Specifications

Melting Point -52°C
Density 1.88
Boiling Point 177°C to 180°C
Refractive Index 1.289
Quantity 25 g
Beilstein 1813883
Solubility Information Miscible with chloroform,cyclohexane,ethyl ether,ethyl acetate,heptane,carbon tetrachloride and petroleum ether. Immiscible with water,methanol,toluene,isopropyl alcohol,benzene and acetone.
Formula Weight 671.1
Percent Purity 90%
Grade Technical
Chemical Name or Material Perfluorotri-n-butylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.