missing translation for 'onlineSavingsMsg'
Learn More

Perfluorotributylamine (>80%), TRC

Product Code. 30417004
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30417004 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30417004 Supplier Toronto Research Chemicals Supplier No. TRCP28671010G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Perfluorotributylamine
CAS 311-89-7
Molecular Formula C12 F27 N
Quantity 10 g
Synonym Perfluorotributylamine, 1-Butanamine','1,1,2,2,3,3,4,4,4-nonafluoro-N,N-bis(1,1,2,2,3,3,4,4,4-nonafluorobutyl)-, 1-Butanamine','1,1,2,2,3,3,4,4,4-nonafluoro-N,N-bis(nonafluorobutyl)- (9CI), Tributylamine','heptacosafluoro- (6CI,7CI,8CI), 1,1,2,2,3,3,4,4,4-Nonafluoro-N,N-bis(1,1,2,2,3,3,4,4,4-nonafluorobutyl)-1-butanamine, Afluid E 18, CT-solv 180, EFL 174S, Eftop EF-L 174, FC 43, FC 47, Flourinert FC 40, Fluorinert 43, Fluorinert FC 43, Fluorocarbon FC 43, Fluosol FC 43, Heptacosafluorotributylamine, Mediflor FC 43, Medifluor FC 47, NSC 3501, Oxyferol, PFTBA, Perfluorotri-n-butylamine, Perfluorotributylamine, Tri (n-butyl perfluoro), Tri(perfluorobutyl)amine, Tris(nonafluorobutyl)amine, Tris(perfluorobutyl)amine
InChI Formula InChI=1S/C12F27N/c13-1(14,7(25,26)27)4(19,20)10(34,35)40(11(36,37)5(21,22)2(15,16)8(28,29)30)12(38,39)6(23,24)3(17,18)9(31,32)33
SMILES FC(F)(F)C(F)(F)C(F)(F)C(F)(F)N(C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
IUPAC Name 1,1,2,2,3,3,4,4,4-nonafluoro-N,N-bis(1,1,2,2,3,3,4,4,4-nonafluorobutyl)butan-1-amine
Molecular Weight (g/mol) 671.09
Formula Weight 670.96
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.