missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Perform™ Classic Concentrate Mucasol

Product Code. 17991302 Shop All Brand Products
Change view
Click to view available options
Size:
2 L
5 L
Unit Size:
2 L
5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17991302 2 L 2 L
17971302 5 L 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17991302 Supplier BRAND™ Supplier No. 44103

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Liquid, alkaline high-performance concentrate for immersion and ultrasonic baths.

  • Liquid, alkaline high-performance concentrate for immersion and ultrasonic baths.
  • Universal cleaner for laboratory equipment and instruments made of glass, porcelain, plastic, rubber and metal.
  • Items are cleaned in about 10 to 30 minutes, without mechanical help. Perform™ classic concentrate mucasol replaces chromosulfuric acid, is free from chlorine and caustic alkalis, is non-toxic, nonaggressive and biodegradable.
  • Prolonged exposure of zinc and non-anodized aluminium surfaces should be avoided.
TRUSTED_SUSTAINABILITY

Specifications

Color-coded No
Form Concentrated Liquid
Scent Original
Sterility Non-sterile
Appearance Light Yellow
Autoclavable Not Autoclavable
Color Translucent Yellow
Disposable Limited-use
Filtered Filtered
Flammable No
Size 2 L
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Perform Concentrate Mucasol Liquid Detergent
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : tripotassium orthophosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.